Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDw60 Antibody (UM4D4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311442
Description
CDw60 Monoclonal antibody specifically detects CDw60 in Human samples. It is validated for ELISASpecifications
CDw60 | |
Monoclonal | |
1 mg/ml | |
ELISA 1-5 ug/ml | |
Mouse | |
Bioreactor concentrate | |
RUO | |
Human | |
IgM κ |
ELISA | |
UM4D4 | |
Unconjugated | |
10 mM PBS | |
Human rheumatoid synovial T cell line (ST-1) | |
0.1 mg | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction