Learn More
Description
Host Species: Mouse
Clone: LN1
Isotype: IgM/κ
Species Reactivity: Human. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes
Molecular Weight: 53 and 87 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Specifications
Specifications
| Antigen | CDw75 Ab-1 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LN1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.