Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDw75 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS130P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS130P0 100 μL Concentrated 200μg/mL
MS130P1 500 μL Concentrated 200μg/mL
MS130P1ABX 100 μL Concentrated 1mg/mL
MS130PABX 200 μL Concentrated 1mg/mL
MS130P 1 mL Concentrated 200μg/mL
MS130R1 1 mL Prediluted
MS130R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS130P0 Supplier Epredia Supplier No. MS130P0
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Epredia™ CDw75 Ab-1, Mouse Monoclonal Antibody.

CDw75 is a neuraminidase-sensitive sialoprotein, present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. LN1 reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease. LN1 is shown to be a reliable antibody for ascribing a B-cell phenotype in known lymphoid tissues.

Host Species: Mouse

Clone: LN1

Isotype: IgM/κ

Species Reactivity: Human. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes

Molecular Weight: 53 and 87 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Specifications

Antigen CDw75 Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LN1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes
Quantity 100 μL
Regulatory Status IVD
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.