Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdx1 Antibody (123a), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250160AF700
Description
Cdx1 Monoclonal specifically detects Cdx1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Cdx1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CDX1 | |
Human Cdx1 N-terminal peptide | |
0.1 mL | |
Chromatin Modifiers, Epigenetics | |
1044 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
123a | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Clone 123a has a degree of cross reactivity with a determinant of different molecular weight when applied by WB and IHC in fibroblasts and smooth muscle cells. The cross reacting product does not appear in epithelial cells (or at least is significantly weaker). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction