Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdx1 Mouse anti-Human, Clone: CDX1/1650, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Cdx1 |
---|---|
Clone | CDX1/1650 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Description
Cdx1 Monoclonal specifically detects Cdx1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Cdx1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Epigenetics | |
10 mM PBS with 0.05% BSA | |
1044 | |
IgG1 κ | |
Protein A or G purified |
CDX1/1650 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
caudal type homeo box transcription factor 1, caudal type homeobox 1, caudal type homeobox transcription factor 1, Caudal-type homeobox protein 1, caudal-type homeobox protein CDX1, homeobox protein CDX-1, MGC116915 | |
Recombinant human full-length Cdx1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title