Learn More
Invitrogen™ CDX1 Polyclonal Antibody, DyLight™ 488

Description
CDX1 Polyclonal Antibody for ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | CDX1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.61 mg/mL |
| Conjugate | DyLight 488 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | CDX1 |
| Gene Accession No. | P18111, P47902 |
| Gene Alias | caudal type homeo box 1; caudal type homeo box transcription factor 1; caudal type homeobox 1; caudal type homeobox transcription factor 1; caudal-type homeobox protein 1; caudal-type homeobox protein CDX1; Cdx; CDX1; Cdx-1; Homeobox protein CDX-1 |
| Gene Symbols | CDX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.