Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2/3 Polyclonal Antibody, Cyanine5, Bioss
SDP

Catalog No. 502279777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-227-9777 Supplier Bioss Supplier No. BS9236RCY5
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDX2/3 Polyclonal antibody specifically detects CDX2/3 in Bovine, Chicken, Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CDX2/3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine5
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene CDX2
Gene Accession No. P43241, Q99626
Gene Alias caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CDX2/3
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100127132, 1045, 12591, 374205, 618679, 66019
Target Species Bovine, Chicken, Human, Mouse, Pig, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.