Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Antibody (CDX2/1690), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$246.61 - $554.73

Specifications

Antigen CDX2
Clone CDX2/1690
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25324720
SDP
View Documents
Novus Biologicals
NBP25324720UG
20 μg
Each for $246.61
Only null left
Add to Cart
 
NB253247100
SDP
View Documents
Novus Biologicals
NBP253247100UG
100 μg
Each for $554.73
Only null left
Add to Cart
 
NB253247200
SDP
View Documents
Novus Biologicals
NBP253247200UG
200 μg N/A N/A N/A
Description

Description

CDX2 Monoclonal specifically detects CDX2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

CDX2
0.2 mg/mL
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Unconjugated
Mouse
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
1045
Recombinant human CDX2 protein fragment (aa150-249) (exact sequence is proprietary)
Primary
Store at 4C.
CDX2/1690
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Monoclonal
Purified
RUO
Human
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2
CDX2
IgG2a κ
Protein A or G purified
40 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.