Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDX2 Antibody (CDX2/4394R), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308743H
Description
CDX2 Monoclonal antibody specifically detects CDX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CDX2 | |
Monoclonal | |
HRP | |
caudal type homeo box transcription factor 2, caudal type homeobox 2, Caudal-type homeobox protein 2, CDX-3, CDX3caudal type homeobox transcription factor 2, homeobox protein CDX-2 | |
Recombinant fragments and synthetic peptides from human CDX2 protein (exact sequences are proprietary) (Uniprot: Q99626) | |
100 μg | |
Cancer, Chromatin Research, DNA Repair, Growth and Development, Transcription Factors and Regulators | |
1045 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
CDX2/4394R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction