Learn More
Invitrogen™ CDX2 Polyclonal Antibody

Description
The amino acid sequence used as immunogen for the GATA-4 antibody is 93% homologous in Rhesus Macaque, 87% homologous in Rat, 82% in Mouse, and 81% homologous in Canine.
Specifications
Specifications
| Antigen | CDX2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | CDX2 |
| Gene Accession No. | P43241, Q99626 |
| Gene Alias | caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor |
| Gene Symbols | CDX2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.