Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDX2 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502206489
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6489 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6489 Supplier Fabgennix Inc Supplier No. CDX2212AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDX2 Polyclonal antibody specifically detects CDX2 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen CDX2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CDX2
Gene Accession No. P43241, Q99626
Gene Alias caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor
Host Species Rabbit
Immunogen Mid-Region epitope synthetic peptide taken within amino acid region 100-200 on human CDX2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1045, 12591, 467350, 66019
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.