Learn More
Invitrogen™ CDX4 Monoclonal Antibody (1E9)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA518575
Catalog No. PIMA518575
Only null left
Add to Cart
Description
Description
Peptide Sequence: RKSELAVNLG LSERQVKIWF QNRRAKERKM IKKKISQFEN SGGSVQSDSD SISPGELPNT FFTTPSAVRG FQPIEIQQVI VSE*.
This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDX4
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
O14627, Q07424
CDX4
CDX4 (NP_005184, 202 a.a. ~ 284 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
1E9
Unconjugated
CDX4
caudal type homeo box 4; caudal type homeobox 4; caudal type homeobox transcription factor 4; caudal-type homeobox protein 4; Cdx3; Cdx-3; CDX4; Cdx-4; Homeobox protein CDX-4; RGD1561529
Mouse
Affinity Chromatography
RUO
1046, 12592, 302400
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.