Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103050 Supplier Invitrogen™ Supplier No. PA5103050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDX4.

This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDX4
Gene Accession No. O14627, Q07424
Gene Alias caudal type homeo box 4; caudal type homeobox 4; caudal type homeobox transcription factor 4; caudal-type homeobox protein 4; Cdx3; Cdx-3; CDX4; Cdx-4; Homeobox protein CDX-4; RGD1561529
Gene Symbols CDX4
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDX4(Accession O14627), corresponding to amino acid residues P38-G88.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1046, 12592, 302400
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.