Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563315 Supplier Invitrogen™ Supplier No. PA563315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RAKERKMIKK KISQFENSGG SVQSDSDSIS PGELPNTFFT TPSAVRGFQP IEIQQVIVSE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDX4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDX4
Gene Accession No. O14627
Gene Alias caudal type homeo box 4; caudal type homeobox 4; caudal type homeobox transcription factor 4; caudal-type homeobox protein 4; Cdx3; Cdx-3; CDX4; Cdx-4; Homeobox protein CDX-4; RGD1561529
Gene Symbols CDX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDX4. Recombinant protein control fragment (Product #RP-100064).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.