Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDY2A Monoclonal Antibody (2E2)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524566

Catalog No. PIMA524566


Only null left
Add to Cart

Description

Description

Peptide Sequence: ASTLSDTKNM EIINSTIETL APDSPFDHKK TVSGFQKLEK LDPIAADQQD TVVFKVTEGK LLRDPLSHPG AEQTGIQNKT QMHPLMSQMS GS.

This intronless gene encodes a protein containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. This protein is localized to the nucleus of late spermatids where histone hyperacetylation takes place. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. Two nearly identical copies of this gene are found in a palindromic region on chromosome Y; this record represents the telomeric copy. Chromosome Y also contains a pair of closely related genes in another more telomeric palindrome as well as several related pseudogenes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDY2A
Monoclonal
Unconjugated
CDY2A
CDY; CDY2; CDY2A; CDY2B; chromodomain protein, Y chromosome, 2; chromodomain protein, Y-linked, 2A; testis-specific chromodomain protein Y 2; Y chromosome chromodomain protein 2A
Mouse
Protein A
RUO
9426
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2E2
PBS with no preservative; pH 7.4
Q9Y6F7
CDY2A
CDY2A (NP_004816, 123 a.a. ~ 214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.