Learn More
CDY2B Mouse anti-Human, Clone: OTI8H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810406

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CDY2B encodes a protein containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. This protein is localized to the nucleus of late spermatids where histone hyperacetylation takes place. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. Two nearly identical copies of this gene are found in a palindromic region on chromosome Y; this record represents the centromeric copy. Chromosome Y also contains a pair of closely related genes in another more telomeric palindrome as well as several related pseudogenes.Specifications
CDY2B | |
Monoclonal | |
Unconjugated | |
CDY2B | |
CDY2B | |
Full length human recombit protein of human CDY2B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
OTI8H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
CDY | |
Mouse | |
Affinity Chromatography | |
RUO | |
203611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.