Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550010 Supplier Invitrogen™ Supplier No. PA550010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CDYL2 protein.

CDYL2 (Chromodomain Y Like 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includemethylated histone binding. An important paralog of this gene isCDYL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDYL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDYL2
Gene Accession No. Q8N8U2
Gene Alias 1700029M19Rik; 4930453I21Rik; Cdyl2; CDY-like 2; chromodomain protein, Y chromosome-like 2; chromodomain protein, Y-like 2; chromodomain Y-like protein 2; Gm20194; PCCP1; prostate cancer candidate protein 1, PCCP1
Gene Symbols CDYL2
Host Species Rabbit
Immunogen Synthesized peptide derived from N-terminal of humanCDYL2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124359
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.