Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117098 Supplier Invitrogen™ Supplier No. PA5117098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, U-937, HeLa, Mouse brain, Mouse spleen, Mouse testis, Mouse lung, Rat brain Immunogen sequence: MSKDKRIKSG KQSSTSKLLR DSRGPSVEKL SHRPSDPGKS KGTSHKRKRI NPPLAKPKKG YSGKPSSGGD RATKTVSYRT TPSGLQIMPL KKSQNGMENG DAGSEKDERH FGNGSHQPGL DLNDHVGEQD MGECDVNHAT LAENGLGSAL TNGGLNLHSP VKRKLEAEKD Y.

CDYL2 (Chromodomain Y Like 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includemethylated histone binding. An important paralog of this gene isCDYL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDYL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDYL2
Gene Accession No. Q8N8U2, Q9D5D8
Gene Alias 1700029M19Rik; 4930453I21Rik; Cdyl2; CDY-like 2; chromodomain protein, Y chromosome-like 2; chromodomain protein, Y-like 2; chromodomain Y-like protein 2; Gm20194; PCCP1; prostate cancer candidate protein 1, PCCP1
Gene Symbols CDYL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-230 of human CDYL2 (NP_6895552).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124359, 292044, 75796
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.