Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDYL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $501.50
Specifications
Antigen | CDYL2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15517820
![]() |
Novus Biologicals
NBP15517820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155178
![]() |
Novus Biologicals
NBP155178 |
100 μL |
Each for $501.50
|
|
|||||
Description
CDYL2 Polyclonal specifically detects CDYL2 in Human samples. It is validated for Western Blot.Specifications
CDYL2 | |
Polyclonal | |
Rabbit | |
Q8N8U2 | |
124359 | |
Synthetic peptides corresponding to CDYL2(chromodomain protein, Y-like 2) The peptide sequence was selected from the N terminal of CDYL2 (NP_689555). Peptide sequence NPPLAKPKKGYSGKPSSGGDRATKTVSYRTTPSGLQIMPLKKSQNGMENG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CDY-like 2, chromodomain protein, Y-like 2, chromodomain Y-like protein 2, FLJ38866 | |
CDYL2 | |
IgG | |
57 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title