Learn More
CEA Monoclonal Antibody (26/3/13), Invitrogen™
Description
This antibody recognizes native human CEACAM5 protein expressed in transfected mammalian cells.
Specifications
Specifications
| Antigen | CEA |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | 26/3/13 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Ceacam5 |
| Gene Accession No. | P06731 |
| Gene Alias | 1600029H12Rik; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CEA; CEACAM5; Meconium antigen 100; OTTHUMP00000199033; pregnancy-specific glycoprotein 30; Psg30; sCD66e; soluble CD66e |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.