Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEA Mouse anti-Human, Clone: C66/1291, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14723
Description
Mouse monoclonal antibody raised against full length recombinant human Carcinoembryonic Antigen (CEA).
- High specificity
- High reactivity
Specifications
CEA | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
Protein A/G purification | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence | |
C66/1291 | |
Mouse monoclonal antibody raised against full length recombinant human Carcinoembryonic Antigen (CEA). | |
In 10 mM PBS. | |
Recombinant protein corresponding to full length human Carcinoembryonic Antigen. | |
100 μg | |
Primary | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction