Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEA Antibody (PARLAM 4), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197718JF646
Description
CEA Monoclonal specifically detects CEA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CEA | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| PARLAM 4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Carcino Embryonic Antigen, Carcinoembryonic antigen | |
| Clone PARLAM 4 is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 Ag 14 mouse myeloma cells with spleen cells from a BABL/c mouse immunized with isolated human CEA. The immunogen has been isolated from human colonic carcinoma cells. | |
| 0.1 mL | |
| Most polyclonal CEA antisera show cross-reactivity with related antigens such as biliary glycoprotein (BGP) and non-specific cross-reacting antigen 1/11 (NCA). This antibody does not show cross reactivity, neither with BGP nor with NCA. In immunoblotting the antibody recognizes a single band of 180 kD. | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction