Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEA Rabbit anti-Human, Unconjugated, Clone: C66/1983R, Abnova™

Catalog No. 89337815 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-815 Supplier Abnova Corporation Supplier No. RAB00390
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CEA Monoclonal antibody specifically detects CEA in Human samples. It is validated for IHC-P.

Specifications

Antigen CEA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C66/1983R
Conjugate Unconjugated
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to full length human CEA
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.