Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (103), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289666AF647
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CEACAM1/CD66a | |
Monoclonal | |
Alexa Fluor 647 | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM1/CD66a (NP_001020083.1; Met1-Gly428). | |
0.1 mL | |
Apoptosis, Cancer | |
634 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
103 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction