Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (10H8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CEACAM1/CD66a |
---|---|
Clone | 10H8 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
CEACAM1/CD66a | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
A synthesized peptide derived from Human CEACAM1/CD66a [UniProt P13688] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10H8 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
634 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title