Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (11-1H), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259674PCP
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Rat samples. It is validated for Flow Cytometry.Specifications
CEACAM1/CD66a | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
CEACAM1 | |
Recombinant soluble rat CEACAM1/CD66a-Fc produced in HEK293 cells | |
0.1 mL | |
Apoptosis, Cancer | |
634 | |
Rat | |
Purified |
Flow Cytometry | |
11-1H | |
Flow Cytometry | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross-react with human CEACAM-1, -3. -4, -5, -6, 7 and -8, mouse CEACAM1 and mouse CEACAM2. Does not bind to the N-domain of rat CEACAM1. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction