Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (28T25) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00 - $524.00
Specifications
Antigen | CEACAM1/CD66a |
---|---|
Clone | 28T25 |
Concentration | LYOPH |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CEACAM1/CD66a | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
CEACAM1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
28T25 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer | |
NP_001703 | |
634 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title