Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (28T25), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221997IR
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
CEACAM1/CD66a | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEACAM1 | |
Human recombinant CEACAM1/CD66a | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
28T25 | |
Western Blot, ELISA, Immunohistochemistry | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer | |
634 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction