Learn More
CEACAM1/CD66a Antibody (723629), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6480MFV450
Description
CEACAM1/CD66a Monoclonal antibody specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| CEACAM1/CD66a | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 634 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 723629 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
| Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-1/CD66a, Glu35-Gly428, Accession # P31809 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 |