Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CD66a Mouse anti-Human, Clone: C5-1X, Novus Biologicals™
SDP

Catalog No. p-7201027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019667 0.02 mg
NBP236278 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019667 Supplier Novus Biologicals Supplier No. NBP2362780.02MG

Mouse Monoclonal Antibody

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA, CyTOF-ready.

Specifications

Antigen CEACAM1/CD66a
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Flow Cytometry
Classification Monoclonal
Clone C5-1X
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml, Flow Cytometry 10ug/ml, ELISA 10ug/ml, Immunohistochemistry 10ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, Sandwich ELISA 1:100-1:2000, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen
Gene Symbols CEACAM1
Host Species Mouse
Immunogen recombinant human soluble CEACAM-1-Fc (produced in HEK293 cells)
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 634
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.