Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM1/CD66a Antibody (CC1), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143390PECY7
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Flow Cytometry.Specifications
CEACAM1/CD66a | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CEACAM1 | |
The immunogen for this antibody was CD66a. | |
0.1 mL | |
Primary | |
Mouse | |
IgG1 |
Flow Cytometry | |
CC1 | |
Flow Cytometry | |
antigen CD66, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen | |
Mouse | |
Protein A or G purified | |
Apoptosis, Cancer | |
634.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction