Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM1/CEACAM5 Monoclonal Antibody (4/3/17), Invitrogen™

Catalog No. 01670180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-180 Supplier Thermo Scientific Supplier No. MA117767

Item Discontinued This product is either new or inactive, and cannot be ordered online.

Mouse Monoclonal Antibody

This antibody recognizes native human CEACAM1, 5 protein expressed in transfected mammalian cells.

Over-expression of CEA/CEACAM5 in tumors of epithelial origin is the basis of its wide-spread use as a tumor marker CEACAM1 expression is down-regulated in many tumors indicating a tumor-suppressive function. The anti-tumor effect may be due to inhibition of tumor angiogenesis, possibly by increased secretion of anti-angiogenic molecules from the cells. The function of CEA family members varies widely: they function as cell adhesion molecules, tumor suppressors, regulators of lymphocyte and dendritic cell activation, receptors of Neisseria species and other bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM1/CEACAM5
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 4/3/17
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene CEACAM1
Gene Accession No. P06731, P13688
Gene Alias antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 5; CD66a; CD66a antigen; CD66e; CEA; CEACAM1; CEACAM5; Meconium antigen 100; pan CEACAM; sCD66e; soluble CD66e
Gene Symbols CEACAM1, CEACAM5
Host Species Mouse
Immunogen Extracted protein of CEACAM5
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048, 634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.