Learn More
CEACAM1 Recombinant Rabbit Monoclonal Antibody (BLR032F), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $595.98
Specifications
Antigen | CEACAM1 |
---|---|
Clone | BLR032F |
Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2658
![]() |
Bethyl Laboratories, Inc
A700032T |
10 μL |
Each for $157.08
|
|
|||||
50-157-2657
![]() |
Bethyl Laboratories, Inc
A700032 |
100 μL |
Each for $595.98
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommende
CEACAM1 is a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregcy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of only two has been determined.Specifications
CEACAM1 | |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P13688 | |
634 | |
recombit protein representing the extracellular domain of human CEACAM1. | |
Primary | |
4° C | |
CEACAM1 |
BLR032F | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
BGP, BGP1, BGPI, CD66a antigen, antigen CD66, carcinoembryonic antigen-related cell adhesion molecule 1 | |
CEACAM1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.