Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM16 Antibody (2747B), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9885F
Description
CEACAM16 Monoclonal antibody specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
CEACAM16 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
388551 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry, Immunocytochemistry | |
2747B | |
Flow Cytometry, Immunocytochemistry | |
carcinoembryonic antigen-related cell adhesion molecule 16 | |
Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction