Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEACAM16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560917 Supplier Invitrogen™ Supplier No. PA560917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPVPTKPTEG QDVTLTVQGY PKDLLVYAWY RGPASEPNRL LSQLPSGTWI AGPAHTGREV GFPNCSLLVQ KLNLTDTGRY TLKTVTVQGK T Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Required for proper hearing, it may play a role in maintaining the integrity of the tectorial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM16
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEACAM16
Gene Accession No. Q2WEN9
Gene Alias B-cell leukemia/lymphoma 3; Bcl3; carcinoembryonic antigen like-2 protein; carcinoembryonic antigen related cell adhesion molecule 16; Carcinoembryonic antigen-like 2; Carcinoembryonic antigen-related cell adhesion molecule 16; Ceacam16; CEAL2; CEA-related cell adhesion molecule 16; DFNA4B; Gm769
Gene Symbols CEACAM16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEACAM16. Recombinant protein control fragment (Product #RP-99226).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388551
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.