Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CEACAM16 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CEACAM16 Polyclonal specifically detects CEACAM16 in Human samples. It is validated for Western Blot.Specifications
CEACAM16 | |
Polyclonal | |
Rabbit | |
carcinoembryonic antigen-related cell adhesion molecule 16 | |
CEACAM16 | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
388551 | |
IgG | |
53 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title