Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM19 Antibody (Z5P2D9*A9), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253124AF647
Description
CEACAM19 Monoclonal specifically detects CEACAM19 in Human samples. It is validated for Western Blot, ELISA.Specifications
CEACAM19 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEACAM19 | |
Peptide sequence - RSINPARPLP | |
0.1 mL | |
Apoptosis, Cancer | |
56971 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
Z5P2D9*A9 | |
Western Blot, ELISA | |
carcinoembryonic antigen-related cell adhesion molecule 19 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction