Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM19 Antibody (Z5P2D9*A9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP253124AF750

Catalog No. NP23124A70


Only null left
Add to Cart

Description

Description

CEACAM19 Monoclonal specifically detects CEACAM19 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CEACAM19
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CEACAM19
Peptide sequence - RSINPARPLP
0.1 mL
Primary
Human
IgG
Western Blot, ELISA
Z5P2D9*A9
Western Blot, ELISA
carcinoembryonic antigen-related cell adhesion molecule 19
Mouse
Protein A purified
Apoptosis, Cancer
56971
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.