Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM2 Antibody (935610), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB9214H
Description
CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CEACAM2 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 935610 | |
| Flow Cytometry, CyTOF-ready | |
| Bgp;Bgp2;Carcinoembryonic antigen-related cell adhesion molecule 2;Ceacam2-L | |
| Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422, Accession # Q925P2 | |
| RUO | |
| 26367 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction