Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM20 Antibody (1032339), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9416MFV500
Description
CEACAM20 Monoclonal antibody specifically detects CEACAM20 in Human samples. It is validated for Flow CytometrySpecifications
CEACAM20 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
1032339 | |
Flow Cytometry | |
UNQ9366 | |
Human embryonic kidney cell HEK293-derived human CEACAM-20, Gln31-Gly450, Accession # Q6UY09-1 | |
RUO | |
125931 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction