Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570882
Description
CEACAM20 Polyclonal Antibody for Western Blot
Together with the tyrosine-protein kinase SYK, enhances production of the cytokine CXCL8/IL-8 via the NFKB pathway and may thus have a role in the intestinal immune response.
Specifications
CEACAM20 | |
Polyclonal | |
Unconjugated | |
CEACAM20 | |
9130012D09Rik; carcinoembryonic antigen related cell adhesion molecule 20; carcinoembryonic antigen-related cell adhesion molecule 20; Ceacam20; CEA-related cell adhesion molecule 20; GPAD9366; UNQ9366; UNQ9366/PRO34155 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
125931 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UY09 | |
CEACAM20 | |
synthetic peptide directed towards the C-terminal region of human CEACAM20. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction