Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM21 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297874V
Description
CEACAM21 Polyclonal specifically detects CEACAM21 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CEACAM21 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
carcinoembryonic antigen-related cell adhesion molecule 21, CEACAM3, FLJ13540, MGC119874, R29124_1 | |
Produced in rabbits immunized with E. coli-derived Human CEACAM21 fragment. | |
0.1 mL | |
Cancer | |
90273 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction