Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEACAM21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101499 Supplier Invitrogen™ Supplier No. PA5101499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEACAM21.

CEACAM21 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 21) is a Protein Coding gene. An important paralog of this gene is CEACAM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM21
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEACAM21
Gene Accession No. Q3KPI0
Gene Alias carcinoembryonic antigen related cell adhesion molecule 21; carcinoembryonic antigen-related cell adhesion molecule 21; CEACAM21; CEACAM3; R29124_1; UNQ3098/PRO10075
Gene Symbols CEACAM21
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEACAM21(Accession Q3KPI0), corresponding to amino acid residues L246-S293.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90273
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.