Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM21 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA532014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532014 Supplier Thermo Scientific Supplier No. PA532014

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CEACAM21 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 21) is a Protein Coding gene. An important paralog of this gene is CEACAM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM21
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CEACAM21
Gene Accession No. Q3KPI0
Gene Alias carcinoembryonic antigen related cell adhesion molecule 21; carcinoembryonic antigen-related cell adhesion molecule 21; CEACAM21; CEACAM3; R29124_1; UNQ3098/PRO10075
Gene Symbols CEACAM21
Host Species Rabbit
Immunogen Recombinant protein fragment corresponding to a region within amino acids 6 and 185 of Human CEACAM21
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.