Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM21 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988194
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8194 Supplier Bioss Supplier No. BS7996R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CEACAM21 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 21) is a Protein Coding gene. An important paralog of this gene is CEACAM1.

Specifications

Antigen CEACAM21
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEACAM21
Gene Accession No. Q3KPI0
Gene Alias carcinoembryonic antigen related cell adhesion molecule 21; Carcinoembryonic antigen-related cell adhesion molecule 21; CEACAM21; CEACAM3; R29124_1; UNQ3098/PRO10075
Gene Symbols CEACAM21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CEACAM21.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90273
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.