Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM3/CD66d Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299909MFV450
Description
CEACAM3/CD66d Polyclonal antibody specifically detects CEACAM3/CD66d in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CEACAM3/CD66d | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282 | |
Produced in rabbits immunized with purified, recombinant Human CEACAM3/CD66d (Accession#: NP_001806.2; Met1-Gly155) | |
0.1 mL | |
CD Markers, Immunology, Innate Immunity | |
1084 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction