Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM3 Monoclonal Antibody (4F1H8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548420
Description
CEACAM3 Monoclonal Antibody for Western Blot, Flow, ELISA
This gene encodes a member of the family of carcinoembryonic antigen-related cell adhesion molecules (CEACAMs), which are used by several bacterial pathogens to bind and invade host cells. The encoded transmembrane protein directs phagocytosis of several bacterial species that is dependent on the small GTPase Rac. It is thought to serve an important role in controlling human-specific pathogens by the innate immune system. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
CEACAM3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P40198, Q63111 | |
CEACAM3 | |
Purified recombinant fragment of human CEACAM3 (AA: 35-155) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot, Immunohistochemistry, Western Blot | |
4F1H8 | |
Unconjugated | |
CEACAM3 | |
carcinoembryonic antigen CGM1; Carcinoembryonic antigen gene family (CGM1); carcinoembryonic antigen gene family 4; carcinoembryonic antigen gene family member 1; Carcinoembryonic antigen gene family member 4 / (carcinoembryonic antigene-related protein); carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen-related cell adhesion molecule 3; CD66d; CD66d antigen; CEA; cea12; CEACAM3; ceacam3 {ECO:0000312; Ceacam5; Cear; Cgm1; Cgm4; CGM4AA; EG384557; nonspecific cross-reacting antigen; pregnancy-specific glycoprotein 24; Psg24; RATCEAA; RATCGM4AA; RGD:2333}; W264; W282 | |
Mouse | |
Protein G | |
RUO | |
1084, 24926 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction