Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM4 Antibody (822608), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7845PECY7
Description
CEACAM4 Monoclonal antibody specifically detects CEACAM4 in Human samples. It is validated for Flow CytometrySpecifications
CEACAM4 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1089 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
822608 | |
Flow Cytometry | |
carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CGM7, CGM7_HUMAN | |
E. coli-derived recombinant human CEACAM-4, Phe36-Val140, Accession # O75871 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction