Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEACAM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113805 Supplier Invitrogen™ Supplier No. PA5113805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTIEALPSSA AEGKDVLLLA CNISETIQAY YWHKGKTAEG SPLIAGYITD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

CEACAM4 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 4) is a Protein Coding gene. An important paralog of this gene is CEACAM3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CEACAM4
Gene Accession No. O75871
Gene Alias carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 10; carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; C-CAM4 protein; Ceacam10; CEACAM4; CEA-related cell adhesion molecule 10; CGM7; CGM7_HUMAN; NCA; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236
Gene Symbols CEACAM4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human CEACAM4 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1089
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.