Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CEACAM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587599
Description
Immunogen sequence: FTIEALPSSA AEGKDVLLLA CNISETIQAY YWHKGKTAEG SPLIAGYITD IQANIPGAAY SGRETVYPNG SLLFQNITLE DAGSYTLRTI NASYDSDQAT GQLHVHQNNV PGLPVGAVAG; Positive Samples: U-87MG, A-549; Cellular Location: Membrane, Single-pass type I membrane protein.
CEACAM4 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 4) is a Protein Coding gene. An important paralog of this gene is CEACAM3.
Specifications
| CEACAM4 | |
| Polyclonal | |
| Unconjugated | |
| CEACAM4 | |
| carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 10; carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; C-CAM4 protein; Ceacam10; CEACAM4; CEA-related cell adhesion molecule 10; CGM7; CGM7_HUMAN; NCA; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1089 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.69 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75871 | |
| CEACAM4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 36-155 of human CEACAM4 (NP_0018082). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction